Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Pharmingen™ Hoechst 34580 Reagent
Shop All BD Biosciences Products
Click to view available options
Quantity:
1 mg
Description
- excitation: UV or violet
- emission: 368nm
- Reconstitue with 1mL distilled water to yield a 1mg/mL solution
- Ribonuclease treatment is not needed to avoid nonspecific RNA staining
Specifications
Specifications
| Content And Storage | -20°C. Protect from light. |
| For Use With (Application) | Flow Cytometry, Fluorescence microscopy |
| Quantity | 1 mg |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction