Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosflow Perm Buffer III, BD
Shop All BD Biosciences Products
Click to view available options
Quantity:
125 mL
Description
- Designed for optimal fluorescent staining in flow cytometry
- Can be used with antibodies for phospho-protein analysis
- In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)
Specifications
Specifications
| Product Type | Staining Buffer |
| Validated Application | RUO |
| Quantity | 125 mL |
| Color | Clear |
| Content And Storage | Room Temperature |
| Format | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction