Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Port-A-Cul™ Anaerobic Collection and Transport Systems

Catalog No. b21608 Shop All BD Diagnostic Systems Products
Click to view available options
Product Type:
Specimen Vial Transport
Transport Container
Transport Tube
Sterility:
Non-sterile
Sterile
This item is not returnable. View return policy
This item is not returnable. View return policy

Versatile—use for fluid, tissue, or swab specimens

  • Collect and maintain specimens at 20° to 25°C for up to 72 hours
  • Ensure aerobic, anaerobic, microaerophilic or facultative microorganism viability during transport from patient to laboratory (including Campylobacter jejuni)
  • Reduced medium formulated to protect oxygen-sensitive microbes
  • Media contain an indicator that continuously monitors the reduced media status
  • Collect specimen on a swab, as a fluid, or as a tissue specimen
  • Available as tube system (capacity 11mL), jar system (capacity 20mL), or vial system (capacity 5mL)
  • Sterile Package contains either one tube and two polyester-tipped wooden swabs, one vial or one jar
  • Screw-cap jar is 3 in. tall and has a 1 in.-wide mouth; not for use with fluids
  • Fluid Collection Kit contains one sterile-packaged vial containing media and a 5cc syringe with a 22-gauge needle and Luer-Lok™ closure

TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Vial Transport
Sterility Non-sterile
For Use With (Application) To maintain the viability of anaerobic, facultative and aerobic microorganisms during transport from the patient to the laboratory
Quantity 10 Pk.
Storage Requirements 20°C to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.