Learn More
Description
Includes
- 558517
- CD3 APC, CD4 PE, CD8 FITC.
- 558512
- CD3 APC, CD25 PE, RT1B FITC.
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC/APC/PE |
| Host Species | Rat |
| Purification Method | Affinity Purified |
| Quantity | 20 Tests |
| Regulatory Status | RUO |
| Includes | CD3 APC, CD4 PE, CD8 FITC |
| Content And Storage | Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Isotype | IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.