Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Rat C-reactive protein (CRP) ELISA Kit
Shop All BD Biosciences Products
Click to view available options
Quantity:
1 Plate
Description
Includes
Capture antibody coated microplate, detection antibody/enzyme conjugate (100x), standard (10x), wash buffer, TMB substrate, stop solution
The C-reactive protein is an acute-phase protein produced by liver during conditions of inflammation, bacterial infection, or tissue trauma
- Reactivity: Rat
- Useful for determination of inflammatory conditions
- Utilizes an antibody specific for rat CRP coated on a 96-well plate
Enzyme-linked immunosorbent assay

Specifications
Specifications
Antigen | CRP |
Cross Reactivity | Rat |
Includes | Rat C-Reactive protein ELISA microplate (96-well coated with Anti-CRP), Rat C-Reactive protein standard (10X), HRP Conjugated Rabbit Anti-rat CRP (100X), PBS-T Wash Buffer powder, TMB Substrate, stop solution |
Quantity | 1 Plate |
Sample Type | Rat Serum |
Content And Storage | 2°C to 8°C |
For Use With (Application) | Detection and quantitation of Rat CRP in Rat Serum, ELISA |
Product Type | Rat C-reactive Protein ELISA kit |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction