Learn More
Description
Includes
Ratiometric Calcium Indicator, lyophilized 10X Signal Enhancer
- For homogeneous measurement of intracellular calcium changes caused by activation of G-protein coupled receptors or calcium channels
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Kit allows homogeneous measurement of intracellular calcium changes caused by activation of G-proteins coupled receptors or calcium channels |
| Includes | 1 vial ratiometric calcium indicator, 10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer |
| Quantity | 10 Preps |
| Type | Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.