Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Reagent Set A

Catalog No. bdb550536
Click to view available options
Quantity:
20 Plates

Includes

Substrate reagent A, substrate reagent B, wash concentrate 20X, stop solution, assay diluent, coating buffer

Use in BD OptEIA™ ELISA Sets

Enzyme-linked immunosorbent assay

TRUSTED_SUSTAINABILITY

Specifications

Product Type OptEIA™ reagent set A
Quantity 20 Plates
For Use With (Application) ELISA
Form ELISA Set
Content And Storage 2°C to 8°C
Kit Contents 250mL Coating Buffer pH 6.5, 1L assay diluent, 1L Wash Concentrate 20X, 125mL Substrate reagent A and 125mL reagent B, 125mL sto p solution

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.