Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human Interleukin-10 (IL-10)

Catalog No. bdb554611 Shop All BD Biosciences Products
Click to view available options
Quantity:
5 μg

Pure by SDS-PAGE analysis, and an absorbance assay based on the Beers-Lambert law

Frozen, sterile-filtered aqueous buffered solution containing glycerol and biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Human
  • Bioassay: The activity range is measured in a proliferation assay using MC/9 indicator cells
  • Specific Activity: 0.5 - 3.3 x 107 Units/mg
  • ED50: 0.3 to 2.0ng/ml
  • Observed dose response: >250 fold

Bioassay, enzyme-linked immunosorbent assay standard, intracellular block/flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Bioassay, ELISA Standard, Intracellular Block/Flow Cytometry
Molecular Weight (g/mol) 18.5
Name Human IP-10 Recombinant Protein
Purification Method SDS-PAGE
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.