Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Recombinant Human Interleukin-12 (IL-12), p40
Click to view available options
Quantity:
5 μg
Description
Pure detemined by SDS-PAGE, and an absorbance assay based on the Beers-Lambert law
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
Enzyme-linked immunosorbent assay standard

Specifications
Specifications
Concentration | 100μg/mL |
For Use With (Application) | ELISA |
Formulation | Frozen aqueous Buffered solution containing BSA |
Name | Human IL-12 (p40) Recombinant Protein |
Purification Method | SDS-PAGE |
Quantity | 5 μg |
Storage Requirements | -80°C |
Cross Reactivity | Human |
Recombinant | Recombinant |
Form | Solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction