Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human Interleukin-7 (IL-7)

Catalog No. bdb554608
Click to view available options
Quantity:
5 μ g

Potent lymphoid cell growth factor which affects pre-B, pro-B, and early T cells

Lyophilized from a solution comprised of 0.22 ++m sterile-filtered PBS and containing 50μg bovine serum albumin per ++g of cytokine

  • Reactivity: Human
  • Storage Buffer: Lyophilized in an aqueous buffered solution containing BSA

Enzyme-linked immunosorbent assay standard, bioassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 5 μ g
Source Human

Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.