Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Recombinant Human Lymphotoxin (LT-α)
Shop All BD Biosciences Products
Click to view available options
Quantity:
10 μg
Description
Potent multifunctional cytokine which is primarily produced by activated lymphocytes. LT-α has cytotoxic activity against a wide range of tumor cells and certain other target cells
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity: Bioactivity is tested in MTT/actinomycin D cytolysis assay using L929 indicator cells
- Specific Activity Range: 0.3 - 5.0x109 Unit/mg
- ED50: 2 - 30 pg/ml
- Observed dose response: > 500 fold
Bioassay, enzyme-linked immunosorbent assay standard

Specifications
Specifications
Concentration | 200μg/mL |
For Use With (Application) | Bioassay, ELISA Standard |
Formulation | Frozen aqueous Buffered solution containing BSA |
Name | Human LT-a Recombinant Protein |
Purification Method | SDS-PAGE |
Quantity | 10 μg |
Storage Requirements | -80°C |
Cross Reactivity | Human |
Recombinant | Recombinant |
Form | Solution |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction