Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Recombinant Mouse nterleukin-12 (IL-12), p40
Shop All BD Biosciences Products
Click to view available options
Quantity:
2 μg
Description
- Storage Buffer: Frozen aqueous buffered solution containing BSA and glycerol
Enzyme-linked immunosorbent assay

Specifications
Specifications
Concentration | 40μg/mL |
For Use With (Application) | Bioassay, ELISA Standard |
Formulation | Frozen aqueous Buffered solution containing BSA and Glycerol |
Name | Mouse IL-12 Recombinant Protein |
Purification Method | SDS-PAGE |
Quantity | 2 μg |
Storage Requirements | -80°C |
Cross Reactivity | Mouse |
Recombinant | Recombinant |
Form | Solution |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction