Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Rat Tumor Necrosis Factor (TNF-α)

Catalog No. bdb555109 Shop All BD Biosciences Products
Click to view available options
Concentration:
100μg/mL

Potent multifunctional cytokine

  • Molecular weight: 17.4kD protein
  • Contains 157 amino acid residues
  • Reactivity: rat
  • Storage buffer: frozen aqueous buffered solution containing BSA
  • Exerts regulatory and cytotoxic effects on normal lymphoid and non-lymphoid cells and tumor cells

Bioassay, ELISA standard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100μg/mL
For Use With (Application) Bioassay, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA
Name TNF-alpha Recombinant Protein
Quantity 5μg
Storage Requirements -80°C
Cross Reactivity Rat
Recombinant Recombinant

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.