Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
TSA w/Lecithin, Polysorbate 80
Packaging Type:
Single Wrapping, Aseptic Fill
Sterile Double Wrapping Plate
VHP Resistant Triple Wrapping Isolator Pack
Description
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
- Withstands an accidental drop from the height of a bench-top workspace
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Beige |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | TSA w/Lecithin, Polysorbate 80 |
Sterility | Sterile |
For Use With (Application) | Cultivation of fastidious microorganisms and visualization of hemolytic reactions |
Packaging Type | Sterile Double Wrapping Plate |
Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction