Learn More
BD Sedi-Stain™ Concentrated Stain for Urinary Sediment
Click to view available options
Quantity:
12.5 mL
60 mL
Description
- Stabilized modification of the Sternheimer-Mablin urinary stain
- Highly selective formula stains blood cells, casts, and other formed elements in urine sediment in a distinctive fashion that permits rapid and accurate identification
- Sedi-Stain is used directly from the dropper bottle and will remain stable indefinitely
Specifications
Specifications
Type | Concentrated Stain |
For Use With (Application) | Highly selective formula stains Blood cells, casts and other formed elements in Urine sediment in a distinctive fashion that permits rapid and accurate identification |
Quantity | 12.5 mL |