Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Taxo™ Differentiation Strips: Reagent - TB Niacin
Click to view available options
Concentration:
5 μg
Description
- Organisms react with chemical agents impregnated on strip resulting in positive or negative reactions
- Combination of positive and negative reaction aids in the identification of the organism

Specifications
Specifications
Concentration | 5 μg |
Content And Storage | -20°C to +8°C |
Packaging Type | Strips |
Quantity | 1 Pack |
For in vitro Diagnostic use. Follow established laboratory procedures in handling and disposing of infectious materials.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction