Learn More
BD Universal Viral Transport Standard and Combo Kits
Description
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory. System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes a universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory. Medium includes protein for stabilization, antibiotics to minimize bacterial and fungal contamination, and a buffer to maintain a neutral pH.
- Need for refrigerated storage and transport on ice is eliminated, decreasing the prevalence of specimen rejection and eliminating messy specimen transport
- Feature a unique blend of antimicrobics that target and suppress bacterial and fungal contamination and are not harmful to viruses, chlamydiae, mycoplasmas or ureaplasmas
- Clinical pathogens are maintained, while growth of contaminants is inhibited
- Preserves viruses and chlamydiae when stored frozen with the presence of a cryoprotectant
- Provided with labeled capture-cap vials designed for transport of the clinical sample
- System is also supplied as a sample collection kit that comprises a package containing one capture-cap vial of medium and a peel pouch incorporating sterile polyester or nylon flocked specimen collection swabs, with scored shafts for easy breakage
- Capture-cap is designed to secure the shaft of the swab sample to the cap, eliminating use of forceps to remove swab in the laboratory

Specifications
Specifications
Product Type | Universal Viral Transport Vial Standard Kit |
For Use With (Application) | Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory |
Includes | 3mL Vial, two sterile regular Polyester-tipped Plastic swabs, scored shaft |
Quantity | 50 Pk. |
Shaft Material | Plastic |
Tip Material | Polyester |
Tip Type | 2 x Regular |
Volume (Metric) | 3 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.