Learn More
BD Universal Viral Transport Standard and Combo Kits
Catalog No. 2203116
Click to view available options
Tip Type:
2 x Regular
Minitip
This item is not returnable. View return policy
This item is not returnable. View return policy
Sterile kits provide room temperature (2° to 25°C) storage and transport of viruses, chlamydiae, mycoplasmas and ureaplasmas
For the collection and transport of clinical specimens containing viruses, chlamydiae, mycoplasmas or ureaplasmas from the collection site to the testing laboratory. System can be processed using standard clinical laboratory operating procedures for viral, chlamydial, mycoplasmal and ureaplasmal culture. Includes a universal transporting medium that is room temperature stable, which can sustain viability (and infectivity) of a plurality of organisms during transit to the testing laboratory. Medium includes protein for stabilization, antibiotics to minimize bacterial and fungal contamination, and a buffer to maintain a neutral pH.
  • Need for refrigerated storage and transport on ice is eliminated, decreasing the prevalence of specimen rejection and eliminating messy specimen transport
  • Feature a unique blend of antimicrobics that target and suppress bacterial and fungal contamination and are not harmful to viruses, chlamydiae, mycoplasmas or ureaplasmas
  • Clinical pathogens are maintained, while growth of contaminants is inhibited
  • Preserves viruses and chlamydiae when stored frozen with the presence of a cryoprotectant
  • Provided with labeled capture-cap vials designed for transport of the clinical sample
  • System is also supplied as a sample collection kit that comprises a package containing one capture-cap vial of medium and a peel pouch incorporating sterile polyester or nylon flocked specimen collection swabs, with scored shafts for easy breakage
  • Capture-cap is designed to secure the shaft of the swab sample to the cap, eliminating use of forceps to remove swab in the laboratory
TRUSTED_SUSTAINABILITY
Specifications
Product Type Universal Viral Transport Combo Kit
For Use With (Application) Collection and transport of clinical specimens containing Viruses, Chlamydiae, Mycoplasmas or Ureaplasmas from the collection site to the testing laboratory
Includes 3mL Vial, one sterile minitip or wire regular Polyester tipped swab, scored Plastic shaft
Quantity 50 Pk.
Shaft Material Plastic
Tip Material Polyester
Tip Type Minitip
Volume (Metric) 3 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.