Learn More
BD Urine Preservative Transport
Catalog No. 22370173
Click to view available options
Additive:
NAP Guard™, 50μL
Includes
Each Qx UPT Kit contains 100 Qx UPT tubes and 100 transfer pipettes. Each UPT contains 50μL of NAP Guard™ (≥ 742.5mM K2EDTA).
Extends range of time and temperature conditions for CT/GC specimen storage and transport
Qx Urine Preservative Transport
  • Allows for an extended range of time and temperature conditions for specimen storage and transport
  • Designed to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae
  • For male and female urine specimens from symptomatic and asymptomatic individuals
  • For use with the BD ProbeTec™ CT Qx and GC Qx Amplified DNA Assays on the BD Viper™ System in extracted mode
TRUSTED_SUSTAINABILITY
Specifications
Product Type Urine Preservative Tube
Additive NAP Guard™, 50μL
For Use With (Application) For BD ProbeTec™ Chlamydia trachomatis and Neisseria gonorrhoeae Qx amplified DNA Assays, to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae in male and female Urine specimens
Includes Each Qx UPT kit contains 100 Qx UPT tubes and 100 transfer pipettes, each UPT contains 50μL of NAP Guard™
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.