Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Vacutainer™ Plastic Blood Collection Tubes with Fluoride: Conventional Stopper

Catalog No. 02683135
Click to view available options
Closure Color:
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy

Evacuated blood collection tubes for glucose determinations

BD Vacutainer™ fluoride tubes are used to collect samples for glucose determinations

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are gray

Plastic Evacuated Tubes

  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation

TRUSTED_SUSTAINABILITY

Specifications

Additive Sodium Fluoride (10 mg), Potassium Oxalate (8 mg)
Material Plastic
Closure Color Gray
Tube Size 13 x 75 mm
For Use With (Application) Glucose determinations
Label Type Paper
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.