Learn More
BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper
Shop All BD Diagnostic Systems ProductsDescription
- Sterile vacuum blood collection tubes for serum determination
- Spray-coated silica interior
- Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
- With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
- Blood is drawn directly from the vein into the evacuated sterile collection tubes
- Only one venipuncture is needed to draw multiple samples
- Tubes can be changed while needle and plastic holder remain in place on patient's arm
- Closures are color coded
- Made of polyethylene terephthalate (PET)
- Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
- In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Specifications
Specifications
| Product Type | Blood Collection Tube |
| Additive | Clot Activator, Silicone Coated |
| Material | Plastic |
| Closure Type | Traditional Cap |
| Capacity (Metric) | 6 mL |
| Closure Color | Red |
| Diameter (Metric) Outer | 13 x 100 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.