Learn More
BD Vacutainer™ Winged Safety Push Button Blood Collection Sets without Luer Adapter
Catalog No. bd367323 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
23 G
25 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
BD367323 25 G
BD367324 23 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BD367323 Supplier BD Supplier No. 367323
This item is not returnable. View return policy
Blood collection set offers in-vein safety activation provides split-second protection against accidental needlestick injuries
  • Ease of use reduces training requirements and improves activation compliance
  • In-vein activation reduces the risk exposure to a contaminated needle
  • Easy activation without additional patient discomfort
  • Ideal for use in high-risk environments No luer adapter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push Button Blood Collection Set
Needle Gauge 25 G
Length (English) Needle 3/4 in.
Length (English) Tube 12 in.