Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDCA-2 (CD303) Mouse anti-Human, Brilliant Violet 605, Clone: V24-785, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748004
Description
BDCA-2 (CD303) Monoclonal antibody specifically detects BDCA-2 (CD303) in Human samples. It is validated for Flow Cytometry.
Specifications
BDCA-2 (CD303) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
BDCA-2 (CD303) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
V24-785 | |
Brilliant Violet 605 | |
50 μg | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction