Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDCA-2 (CD303) Mouse anti-Human, BV421, Clone: V24-785, BD Horizon™
Mouse Monoclonal Antibody
$225.00 - $416.00
Specifications
Antigen | BDCA-2 (CD303) |
---|---|
Clone | V24-785 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
BDCA-2 (CD303) Monoclonal antibody specifically detects BDCA-2 (CD303) in Human samples. It is validated for Flow Cytometry.Specifications
BDCA-2 (CD303) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
Primary |
V24-785 | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title