Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDCA-2 (CD303) Mouse, anti-Human, R718, Clone: V24-785, BD OptiBuild™

Catalog No. BDB751950 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751950 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751950 Supplier BD Biosciences Supplier No. 751950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The V24-785 monoclonal antibody binds specifically to BDCA-2 (Blood Dendritic Cell Antigen 2), also known as CD303. As a member of the C-type lectin (CLEC) superfamily, it is an adhesion receptor that binds to galactose-terminated desialylated glycans on cell surfaces and on serum IgG. In the blood, BDCA-2 is expressed only on plasmacytoid dendritic cells (pDC). TLR-9 mediates the production of large amounts of IFN-α and IFN-β by activated pDC, and crosslinking of BDCA-2 inhibits that response.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BDCA-2 (CD303)
Applications Flow Cytometry
Classification Monoclonal
Clone V24-785
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.