Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BDH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518866
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518866 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518866 Supplier Invitrogen™ Supplier No. PA518866
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

Dehydrogenase that mediates the formation of 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin and associates with LCN2, thereby playing a key role in iron assimilation and homeostasis. Plays a role in susceptibility to bacterial infection by providing an assimilable source of iron that is exploited by pathogenic bacteria. Also acts as a 3-hydroxybutyrate dehydrogenase (PubMed:16380372). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BDH2
Gene Accession No. Q9BUT1
Gene Alias 1810026B04Rik; 3-hydroxybutyrate dehydrogenase 2; 3-hydroxybutyrate dehydrogenase type 2; 3-hydroxybutyrate dehydrogenase, type 2; bdh2; dehydrogenase/reductase (SDR family) member 6; dehydrogenase/reductase SDR family member 6; Dhrs6; EFA6R; FLJ13261; hypothetical protein LOC550507; oxidoreductase UCPA; PRO20933; R-beta-hydroxybutyrate dehydrogenase; SDR15C1; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1; si:dkey-162b23.2; UCPA-OR; UNQ6308; UNQ6308/PRO20933; zgc:110323
Gene Symbols BDH2
Host Species Goat
Immunogen Synthetic peptide sequence (TKKKQIDQFANE) corresponding to the internal amino acids of BDH2 (aa 60-72).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56898
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.