Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BDH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557501 Supplier Invitrogen™ Supplier No. PA557501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGAKVIATDI NESKLQELEK YPGIQTRVLD VTKKKQIDQF ANEVERLDVL FNVAGFVHHG TVLDCEEKDW DFS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.

Dehydrogenase that mediates the formation of 2,5-dihydroxybenzoic acid (2,5-DHBA), a siderophore that shares structural similarities with bacterial enterobactin and associates with LCN2, thereby playing a key role in iron assimilation and homeostasis. Plays a role in susceptibility to bacterial infection by providing an assimilable source of iron that is exploited by pathogenic bacteria. Also acts as a 3-hydroxybutyrate dehydrogenase (PubMed:16380372). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDH2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BDH2
Gene Accession No. Q9BUT1
Gene Alias 1810026B04Rik; 3-hydroxybutyrate dehydrogenase 2; 3-hydroxybutyrate dehydrogenase type 2; 3-hydroxybutyrate dehydrogenase, type 2; bdh2; dehydrogenase/reductase (SDR family) member 6; dehydrogenase/reductase SDR family member 6; Dhrs6; EFA6R; FLJ13261; hypothetical protein LOC550507; oxidoreductase UCPA; PRO20933; R-beta-hydroxybutyrate dehydrogenase; SDR15C1; Short chain dehydrogenase/reductase family 15C member 1; short chain dehydrogenase/reductase family 15C, member 1; si:dkey-162b23.2; UCPA-OR; UNQ6308; UNQ6308/PRO20933; zgc:110323
Gene Symbols BDH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BDH2. Recombinant protein control fragment (Product #RP-96716).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56898
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.