Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDKRB1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8701 Supplier Bioss Supplier No. BS8675R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.

Specifications

Antigen BDKRB1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Bdkrb1
Gene Accession No. P46663
Gene Alias B1 bradykinin receptor; B1bkr; B1R; Bdkrb; Bdkrb1; BDKRB2; Bk b1 receptor; BK-1 receptor; BKB1R; Bkr; BKR1; BKRB1; BRADYB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B1; bradykinin receptor B2; bradykinin receptor, beta 1; KB1; kinin B1; Kinin B1 receptor
Gene Symbols Bdkrb1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BDKRB1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 623
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.