Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BDKRB2 Recombinant Rabbit Monoclonal Antibody (5O4M5)
GREENER_CHOICE

Catalog No. PIMA535573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535573 Supplier Invitrogen™ Supplier No. MA535573
Add to Cart
Add to Cart

BDKRB2 Recombinant Rabbit Monoclonal Antibody (ARC1121)

BDKRB2 Recombinant Monoclonal Antibody for Western Blot

Bradykinin B2 receptor (BDKRB2) expression has been reported in blood, brain, eye, kidney, liver, lung, prostate, skin, small intestine, synovium, uterus, and vessel. ESTs have been isolated from colon, connective tissue, heart, heart/melanocyte/uterus, and kidney libraries. It is associated with G-proteins that activate a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDKRB2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5O4M5
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BDKRB2
Gene Accession No. P25023
Gene Alias B(2); B2; B2 bradykinin receptor; B2BKR; B2BRA; B2R; BDKRB2; BK2; BK-2; BK-2 receptor; BK2R; BK-2R; BK-2Rs; BKR2; bradykinin receptor B2; bradykinin receptor, beta 2; BRB2; DKFZp686O088; kinin B2
Gene Symbols BDKRB2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 302-391 of human BDKRB2 (P30411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25245
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.