Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BDKRB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533362
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset (94%) Mouse, Pig (83%).
Bradykinin B2 receptor (BDKRB2) expression has been reported in blood, brain, eye, kidney, liver, lung, prostate, skin, small intestine, synovium, uterus, and vessel. ESTs have been isolated from colon, connective tissue, heart, heart/melanocyte/uterus, and kidney libraries. It is associated with G-proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications
| BDKRB2 | |
| Polyclonal | |
| Unconjugated | |
| BDKRB2 | |
| B(2); B2; B2 bradykinin receptor; B2BKR; B2BRA; B2R; BDKRB2; BK2; BK-2; BK-2 receptor; BK2R; BK-2R; BK-2Rs; BKR2; bradykinin receptor B2; bradykinin receptor, beta 2; BRB2; DKFZp686O088; kinin B2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12062, 25245, 624 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P25023, P30411, P32299 | |
| BDKRB2 | |
| Synthetic 18 amino acid peptide from C-terminus of human BDKRB2 / BK-2. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction