Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDNF Antibody (010), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290493G
Description
BDNF Monoclonal specifically detects BDNF in Mouse samples. It is validated for Western Blot, ELISA.Specifications
BDNF | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
10 | |
Western Blot, ELISA | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse BDNF (P21237; Met1-Arg249). | |
0.1 mL | |
Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience | |
627 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction