Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDNF Antibody (35928.11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010505AF594
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
BDNF | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
BDNF | |
Recombinant human BDNF (rhBDNF) expressed in Sf 21 insect cells. | |
0.1 mL | |
Primary | |
Monoclonal Anti-Human BDNF will neutralize the biological activity of rhBDNF. This antibody shows < 2% cross-reactivity with rhbeta-NGF, rhNT-3 and rhNT-4 based on ELISA and immunoblotting. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
35928.11 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
Mouse | |
Protein A purified | |
Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience | |
627.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction