Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDNF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 17 publications
$244.50 - $501.50
Specifications
Antigen | BDNF |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB198682SS
![]() |
Novus Biologicals
NB10098682SS |
0.025 mL |
Each for $244.50
|
|
|||||
NB10098682
![]() |
Novus Biologicals
NB10098682 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
BDNF Polyclonal specifically detects BDNF in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
BDNF | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P23560 | |
627 | |
A synthetic peptide from n-terminal region of human mature BDNF conjugated to blue carrier protein was used as the antigen. The peptide is homologous in many other species including rat, mouse, zebra fish and Xenopus species. | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
Specific for mature BDNF. |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:1000 | |
Polyclonal | |
Rabbit | |
Cytokine Research, Immunology, Neuroscience | |
Lyophilized from whole antisera with No Preservative | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
BDNF | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title