Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BDNF Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142306
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise all reported isoforms (NP_001700.2; NP_001700.2; NP_733928.1; NP_733929.1; NP_733930.1; NP_733931.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Specifications
BDNF | |
Polyclonal | |
Unconjugated | |
BDNF | |
Abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
627 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P23560 | |
BDNF | |
Peptide with sequence ETKCNPMGYTKE. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction