Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDNF Rabbit anti-Human, Clone: SJ12-09, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267410
Description
BDNF Monoclonal specifically detects BDNF in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
BDNF | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin | |
Rabbit | |
Protein A purified | |
Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience | |
627 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SJ12-09 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
BDNF | |
Synthetic peptide within C-terminal human BDNF. (SwissProt: P23560 Human; SwissProt: P21237 Mouse; SwissProt: P23363 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction