Learn More
Invitrogen™ BDNF Recombinant Rabbit Monoclonal Antibody (SJ12-09)

Description
BDNF Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | BDNF |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | SJ12-09 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | BDNF |
| Gene Accession No. | P21237, P23363, P23560 |
| Gene Alias | abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.