Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BDP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585567
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585567 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585567 Supplier Invitrogen™ Supplier No. PA585567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (82%), Rat (83%). Positive Control: A. 30 μg 293T whole cell extract, B. 30 μg 293T nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ARID family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ARID3B
Gene Accession No. Q8IVW6
Gene Alias ARID domain-containing protein 3B; ARID3B; AT rich interactive domain 3B (Bright like); AT rich interactive domain 3B (BRIGHT- like); AT rich interactive domain 3B (BRIGHT-like); AT-rich interaction domain 3B; AT-rich interactive domain-containing protein 3B; Bdp; bright and dead ringer gene product homologous protein Bdp; bright and dead ringer protein; Bright-like; Bright-like protein; dead ringer homolog 2; Dri2; DRIL2
Gene Symbols ARID3B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARID3B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10620
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.