Learn More
Description
BDY FL, SE is a fluorescent green BDY (BODIPY™ or boron-dipyrromethene) dye for the labeling of amines. Exhibits similar excitation and emission spectra to dyes TFAX 488 and FITC . Displays high fluorescence quantum yield and high extinction coefficient and is relatively insensitive to pH change. This dye contains a C3 alkyl spacer between the fluorophore and the NHS ester group, reducing the potential for interactions between fluorophore and conjugated biomolecule. The hydrophobic nature of BDY FL makes it ideal for labeling lipids and cell membranes. BDY FL also exhibits narrow emission bandwidths and has a relatively long excited-state lifetime, making it ideal for fluorescence polarization assays and two-photon excitation (TPE) microscopy. Excitation maximum ∽504 nM; emission maximum ∽510 nM.
For more information on how BDY FL, SE may be used, see our protocol: Conjugation Protocol for Amine Reactive Dyes .
BODIPY™ is a registered trademark of Molecular Probes.
Specifications
Specifications
| Chemical Name or Material | (T-4)-[1-[3-[5-[(3,5-Dimethyl-2 H-pyrrol-2-ylidene-κN)methyl]-1 H-pyrrol-2-yl-κN]-1-oxopropoxy]-2,5-pyrrolidinedionato]difluoroboron |
| CAS | 146616-66-2 |
| Quantity | 5 mg |
| Target | Fluorescent Dyes |
| Molecular Formula | C18H18BF2N3O4 |
| Formula Weight | 389.16 |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

