Learn More
Description
BDY FL Staurosporine is a fluorescent kinase-targeting probe. It can be paired with antibodies labelled with TR-FRET donors such as CoraFluor™ 1 (Cat. No. 7920) for high throughput binding and protein quantification assays. BDY FL Staurosporine shows high specificity for tyrosine kinase (TK) and tyrosine kinase-like (TKL) family kinases. Excitation and emission maxima (λ) are 514 and 516 nm, respectively; quantum yield = 0.11; extinction coefficient = 35,000 M-1cm-1.
The measured KD values, from TR-FRET based saturation binding assay data, are provided in the accompanying supplementary file (see below).Specifications
Specifications
| CAS | 2411845-57-1 |
| Quantity | 100 μg |
| Product Line | Tocris |
| Target | Enzyme Probes and Enzyme Substrates |
| Molecular Formula | C49H54BF2N7O7 |
| Purity | 0.95 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

