Learn More
Fisherbrand™ Bead Mill 4 Mini Homogenizer PROMO
Shop All Fisherbrand Products
Click to view available options
:
Bead Mill 4 Mini Homogenizer; Product Type: Bead Mill Homogenizer
Description
- Broad performance range, from gentle mixing to high force homogenization
- Small footprint requires minimal bench space
- Suited for homogenizing up to 4 samples in seconds
- Reproducible extraction of proteins and nucleic acids
- Simultaneously process 4 x 0.5 mL, 4 x 1.5 mL, 4 x 2 mL or 1 x 7 mL samples
Specifications
Specifications
Product Type | Bead Mill Homogenizer |
Includes | Bead Mill 4 Motor Drive, Integrated 0.5, 1.5 and 2 mL Tube Carriage, 7 mL Tube Carriage, Tool Kit, Instruction Manual |