Learn More
Omni International Bead Ruptor 96 Stainless Steel Milling Balls
GREENER_CHOICE
Catalog No. p-7225671 Shop All Omni International Inc Products
Click to view available options
Diameter (Metric):
10 mm
25 mm
This item is not returnable. View return policy
This item is not returnable. View return policy
Stainless Steel Milling Balls must be used with stainless steel jars.
  • Set of Stainless Steel Milling Balls
Specifications
Product Type Milling Balls
Diameter (Metric) 10 mm
Autoclavable Autoclavable
Material Stainless Steel
For Use With (Application) Dry Grinding or Cryomilling
For Use With (Equipment) Stainless Steel Jars
Packaging Quantity 10 Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.