Description
Sturdy polypropylene construction makes these beakers durable, with excellent chemical resistance.
- Polypropylene
- Autoclavable
- Raised graduations
- Large, strong handles with thumb grip for safe handling of acids, alkalis, and other hazardous chemicals.
Specifications
Specifications
| Capacity (Metric) | 3000 mL |
| Material | Polypropylene |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Scale | Embossed Scale |
| Style | Handled |