Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BEAN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562855
Description
Immunogen sequence: RHREYEHGYV SDEHTYSRSS RRMRYACSSS EDWPPPLDIS SDGD Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 68%.
The protein encoded by this gene is one of several proteins that interact with NEDD4, a member of a family of ubiquitin-protein ligases. These proteins have PY motifs in common that bind to the WW domains of NEDD4. NEDD4 is developmentally regulated, and is highly expressed in embryonic tissues. Mutations in this gene (i.e., intronic insertions of >100 copies of pentanucleotide repeats including a (TGGAA)n sequence) are associated with spinocerebellar ataxia type 31. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2010].
Specifications
BEAN | |
Polyclonal | |
Unconjugated | |
BEAN1 | |
BEAN; Bean1; brain expressed, associated with NEDD4, 1; brain-expressed protein associating with Nedd4; Brain-expressed protein associating with Nedd4 homolog; protein BEAN; protein BEAN1; RGD1562003; SCA31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3B7T3 | |
BEAN1 | |
Recombinant protein corresponding to Human BEAN. Recombinant protein control fragment (Product #RP-100438). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction