Learn More
HemoCue America Beckman Coulter™ Gastroccult™ Gastric Sample Occult Blood Test Kit
Catalog No. p-111478
Change view
Click to view available options
Quantity:
40 Pack
40 Tests
6 x 15 mL
Type:
Gastroccult Developer
Occult Blood Test Kit
Straw Applicator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
SK-66040 40 Tests Occult Blood Test Kit
SK-66115 6 x 15 mL Gastroccult Developer
SK-66140 40 Pack Straw Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SK-66040 Supplier HemoCue America Supplier No. 66040A
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Two tests in one
  • CLIA Complexity: Waived
  • CPT Codes: 87723 for occult blood, 83986 for pH
  • Slide test for detecting occult blood and determining pH in gastric samples, both aspirate and vomitus, on the same test card
  • On-slide Performance Monitor assures that the occult blood test is performing to product specifications
  • Scale of pH test ranges from a clinically useful 1.3 to 7.8
  • Both test results are available within one minute
  • Test cards, developer, and applicator straws sold separately
TRUSTED_SUSTAINABILITY
Specifications
CPT Code 87723
Clia Complexity Waived
Detectable Analytes Occult Blood
Type Occult Blood Test Kit
Includes Convenient pH comparison chart, on-slide performance Monitor™, 40 Gastroccult™ Tests and instructions
Quantity 40 Tests
Test Time 1 min.