Learn More
HemoCue America Beckman Coulter™ Hemoccult™ ICT Immunochemical Fecal Occult Blood Test Kit
Description
Includes
ICT FOBT Kit (Cat. No. 23-280-012) includes test devices and 8mL bottle of Hemoccult ICT Buffer; ICT Sample Collection Kit (Cat. No. 23-280-011) contains Patient Kits; each Patient Kit contains Dispensing Envelope with Patient Instructions, Hemoccult ICT Collection Cards, Collection Tissues, Applicator Sticks, and Mailing Pouch; ICT Collection Cards (Cat. No. 23-280-010) contain Collection Cards and Applicator Sticks; ICT Control Kit (Cat. No. 23-280-013) contains bottles (Positive and Negative)
- CLIA Complexity: Waived
- CPT Codes: 82274QW, G0328QW
- Rapid, qualitative immunochemical chromatographic method for detection of human hemoglobin from blood in fecal samples
- Visually read results
Specifications
Specifications
CPT Code | 82274QW, G0328QW |
Clia Complexity | Waived |
Type | ICT Sample Collection Kit |
For Use With (Application) | Detection of Human hemoglobin from Blood in fecal samples |
Includes | Dispensing envelope with patient instructions, Hemoccult ICT collection cards, collection tissues, applicator sticks and mailing pouch |
Quantity | 40 Pack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.