Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beclin 1 Monoclonal Antibody (117CT2.5.1.1)
GREENER_CHOICE

Catalog No. PIMA537470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537470 Supplier Invitrogen™ Supplier No. MA537470
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Beclin 1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 117CT2.5.1.1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.09% sodium azide; pH 7.4
Gene BECN1
Gene Accession No. Q14457
Gene Alias 4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097
Gene Symbols BECN1
Host Species Mouse
Immunogen purified His-tagged recombinant full length human Autophagy BECN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.