Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beclin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-249 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-249 Supplier Invitrogen™ Supplier No. PA116939
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for BECN1 (transient overexpression lysate).

Beclin 1 plays a central role in autophagy. It acts as a core subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate. Different PI3K complex forms are believed to play a role in multiple membrane trafficking pathways - PI3KC3-C1 is involved in the initation of autophagosomes and PI3KC3-C2 is involved in the maturation of autophagosomes and endocytosis. Beclin 1 also plays a role in antiviral host defense.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.1% sodium azide
Gene BECN1
Gene Accession No. O88597, Q14457
Gene Alias 4921513J16Rik; 5430417M23Rik; APG6; Atg6; ATG6 autophagy related 6 homolog; Bcl-2-interacting protein beclin; beclin 1; beclin 1 (coiled-coil, moesin-like BCL2-interacting protein); beclin 1 (coiled-coil, myosin-like BCL2 interacting protein); beclin 1 (coiled-coil, myosin-like BCL2-interacting protein); beclin 1, autophagy related; beclin1; beclin-1; Beclin-1; beclin-1; Beclin-1-C 35 kDa; Beclin-1-C 37 kDa; becn1; Coiled-coil myosin-like BCL2-interacting protein; coiled-coil, myosin-like BCL2-interacting protein; GT197; hypothetical protein MGC73097; myosin-like BCL2 interacting protein; Protein GT197; testis secretory sperm-binding protein Li 215e; VPS30; zgc:73097
Gene Symbols BECN1
Host Species Rabbit
Immunogen Full length human Beclin 1 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56208, 8678
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.