Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclin 2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060982AF350
Description
Beclin 2 Polyclonal specifically detects Beclin 2 in Human samples. It is validated for Western Blot.Specifications
Beclin 2 | |
Polyclonal | |
Western Blot | |
BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 | |
Rabbit | |
0.1 mL | |
441925 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
BECN2 | |
A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot No. A8MW95] | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction