Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beclin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA146312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146312 Supplier Invitrogen™ Supplier No. PA146312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: 293 whole cell lysate, HEK293 kidney cell lysates and MCC827 lung cell lysates. Does not appear to work with A431 lysates.

Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 2
Applications Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.1% sodium azide
Gene Becn2
Gene Accession No. A8MW95, P0DM65
Gene Alias Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein)
Gene Symbols Becn2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 200-300) of the human Beclin 2 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226720, 441925
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.