Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Beclin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572828
Description
Beclin 2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt].
Specifications
Beclin 2 | |
Polyclonal | |
Unconjugated | |
Becn2 | |
Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; beclin 2; beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; BECN1L1; BECN1P1; BECN2; Gm104; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein; RGD1563293; similar to Beclin 1 (Coiled-coil myosin-like BCL2-interacting protein) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A8MW95 | |
Becn2 | |
Beclin 2 antibody for a 16 amino acid peptide near the amino terminus of human Beclin 2. The immunogen is within amino acids 30 - 80 of Beclin 2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction